EMAIL THIS PAGE TO A FRIEND
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society
Label-retaining cells in the rat submandibular gland.
PMID 17875657
Abstract
To identify stem cells in salivary glands, label-retaining cells (LRCs) were established in rat submandibular glands. Developing and regenerating glands were labeled with bromodeoxyuridine (BrdU). To cause gland regeneration, the glands were injured by duct obstruction. BrdU LRCs were observed in all the parenchymal structures except for the acinus of the glands labeled during regeneration. Among these LRCs, a few, but not many, expressed neither keratin18 (K18; an acinar/duct cell marker) nor alpha-smooth muscle actin (alphaSMA; a myoepithelial cell marker), and thus were putative stem cells. These (K18 and alphaSMA)(neg) LRCs were invariably observed in the intercalated duct and the excretory duct. In the intercalated duct, they were at the proximal end bordering the acinus (the neck of the intercalated duct). Next, to test the above identification, gland extirpation experiments were performed. LRCs were established by labeling developing glands with iododeoxyuridine (IdU) in place of BrdU. Removal of one submandibular gland forced the IdU-LRCs in the remaining gland to divide. They were labeled with chlorodeoxyuridine (CldU). The (K18 and alphaSMA)(neg) LRCs in the neck of the intercalated duct and in the excretory duct did not change in number or in IdU label. The CldU label appeared in these cells and then disappeared. These results indicate that the (K18 and alphaSMA)(neg) LRCs have divided asymmetrically and are thus considered salivary gland stem cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6891
5-Chloro-2′-deoxyuridine, thymidine analog
C9H11ClN2O5