EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
In vitro efficacy of new antifolates against trimethoprim-resistant Bacillus anthracis.
PMID 17875993
Abstract
Bacillus anthracis is innately resistant to trimethoprim (TMP), a synthetic antifolate that selectively inhibits several bacterial dihydrofolate reductases (DHFRs) but not human DHFR. Previously, we were able to confirm that TMP resistance in B. anthracis (MIC > 2,048 microg/ml) is due to the lack of selectivity of TMP for the B. anthracis DHFR (E. W. Barrow, P. C. Bourne, and W. W. Barrow, Antimicrob. Agents Chemother. 48:4643-4649, 2004). In this investigation, 24 2,4-diaminopyrimidine derivatives, representing a class of compounds with dihydrophthalazine side chains, were screened for their in vitro effects on B. anthracis Sterne and their selectivities for the B. anthracis DHFR. MICs were obtained by a colorimetric (Alamar blue) broth microdilution assay. Purified human recombinant DHFR (rDHFR) and B. anthracis rDHFR were used in a validated enzyme assay to determine the 50% inhibitory concentrations (IC(50)s) and the selectivity ratios of the derivatives. The MICs ranged from 12.8 to 128 microg/ml for all but nine compounds, for which the MICs were > or =128 microg/ml. The IC(50) values for B. anthracis rDHFR ranged from 46 to 600 nM, whereas the IC(50) values for human rDHFR were >16,000 nM. This is the first report on the in vitro inhibitory actions of this class of antifolates against TMP-resistant B. anthracis isolates. The selective inhibition of B. anthracis rDHFR and the in vitro activity against B. anthracis demonstrate that members of this class of compounds have the potential to be developed into clinically important therapeutic choices for the treatment of infections caused by TMP-resistant bacteria, such as B. anthracis.