EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Pyrrolidine dithiocarbamate and diethyldithiocarbamate are active against growing and nongrowing persister Mycobacterium tuberculosis.
PMID 17876006
Abstract
Diethyldithiocarbamate (DETC) and pyrrolidine dithiocarbamate (PDTC) were highly active against tubercle bacilli, with MICs of 8 microg/ml and 0.13 microg/ml, respectively. DETC and PDTC were active against old cultures, enhanced pyrazinamide or pyrazinamide/rifampin activity, and had serum inhibitory titers of 1:2 and 1:4, respectively, in mice given 100 mg/kg orally.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36650
DCC, puriss., ≥99.0% (GC)
C13H22N2