EMAIL THIS PAGE TO A FRIEND
The Journal of the Oklahoma State Medical Association
Part XI. Trimethoprim-sulfamethoxazole, nitrofurantoin, chloramphenicol, metronidazole and tinidazole, rifaximin, and nitazoxanide.
PMID 17896614