EMAIL THIS PAGE TO A FRIEND
Nihon yakurigaku zasshi. Folia pharmacologica Japonica
[Pharmacological profiles and clinical effects of ropinirole hydrochloride (ReQuip)].
PMID 17938518
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R2530
Ropinirole hydrochloride, powder, ≥98% (HPLC)
C16H24N2O · HCl