EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and biological evaluation of amide derivatives of (5,6-dimethoxy-2,3-dihydro-1H-inden-1-yl)acetic acid as anti-inflammatory agents with reduced gastrointestinal ulcerogenecity.
PMID 17945398
Abstract
A variety of amide derivatives of (5,6-dimethoxy-2,3-dihydro-1H-inden-1-yl)acetic acid were synthesized and screened for their analgesic and anti-inflammatory activities. The compounds were found to have longer activity profile exceeding that of indomethacin in carrageenan-induced rat paw edema model. Few selected compounds were also screened for their antipyretic, anti-arthritic and ulcerogenecity potential. From these studies it can be concluded that these compounds though have significant antipyretic activity did not act through the inhibition of TNF-alpha. The test compounds failed to prevent the development of secondary inflammation in adjuvant-induced arthritis assay. However, these compounds showed no ulcer formation at the tested dose level of 100 mg/kg p.o.