EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Design, synthesis, and melatoninergic activity of new azido- and isothiocyanato-substituted indoles.
PMID 17988084
Abstract
To develop irreversibly binding ligands for the melatonin receptor(s) as tools for tracing the primary melatonin binding site, we report on the design and synthesis of new melatoninergic azido- and isothiocyanato-substituted indoles. All active compounds were partial agonists or antagonists in the Xenopus melanophore assay, the most potent being the 5-OMe C3-substituted azido 45 and isothiocyanato 46 analogues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
687227
1H-Indole-2-methanol, 90%
C9H9NO