EMAIL THIS PAGE TO A FRIEND
International journal of nanomedicine
Pi-Pi complexation of bupivacaine and analogues with aromatic receptors: implications for overdose remediation.
PMID 18019843
Abstract
The interaction of the important but often overdosed local anesthetic bupivacaine, its structural analogs 2,6-dimethylaniline, and N-methyl-2,6-dimethylacetanilide, and cocaine, with several electron deficient aromatic moieties were studied primarily by proton NMR and UV-visible spectroscopy. In solution, the anesthetic, its analogs and cocaine are electron donors and form pi-pi charge transfer complexes with strong aromatic acceptors, as monitored by the upfield changes induced in the NMR chemical shifts (delta) and red-shifted UV-vis wavelength (lamda max) absorbance of the acceptors. The equilibrium binding constant, K, was determined from the 1H NMR charge transfer induced chemical shift changes and used to calculate the free energy (deltaG) for complex formation of three acceptor-donor pairs. HPLC results indicate that the concentrations of free bupivacaine, its analogs and of cocaine are reduced from solution via binding to aromatic-functionalized silica.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
163252
3,4-Dimethoxytoluene, 96%
C9H12O2
184144
3,5-Dinitrobenzyl alcohol, 98%
C7H6N2O5