EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Tetracycline resistance in Ureaplasma spp. and Mycoplasma hominis: prevalence in Bordeaux, France, from 1999 to 2002 and description of two tet(M)-positive isolates of M. hominis susceptible to tetracyclines.
PMID 18025113
Abstract
Twenty-four of 128 clinical isolates of Mycoplasma hominis and 6 of 276 clinical isolates of Ureaplasma spp. from Bordeaux, France (1999 to 2002), were resistant to tetracycline and harbored the tet(M) gene. For M. hominis, we also found an increase in tetracycline resistance and two tet(M)-positive isolates that were susceptible to tetracyclines.