EMAIL THIS PAGE TO A FRIEND
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Fragrance material review on methyl salicylate.
PMID 18031908
Abstract
A toxicologic and dermatologic review of methyl salicylate when used as a fragrance ingredient is presented.