EMAIL THIS PAGE TO A FRIEND
Biulleten' eksperimental'noi biologii i meditsiny
[Antiradical activity of 3-substituted coumarins and their effect on iron-dependent chemiluminescence].
PMID 1804341
Abstract
The antioxidant capacity of 3-aminocoumarin, 3-oxycoumarin, 3-acetylaminocoumarin, and 3-coumarin carbonic acid has been investigated with chemiluminescence measurement and by the accumulation of TBA-active products. All coumarins were found to be antioxidants, with 3-oxy-, 3-amino- and 3-acetylamino coumarins being capable of amplifying chemiluminescence at early stages of the process.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
411183
3-Aminocoumarin, 97%
C9H7NO2