EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Bismesitylmagnesium: a thermally stable and non-nucleophilic carbon-centred base reagent for the efficient preparation of silyl enol ethers.
PMID 18049749
Abstract
Bismesitylmagnesium has been established as an accessible, practical, convenient, and non-nucleophilic carbon-centred base reagent for efficient access to silyl enol ethers from a series of ketone substrates at readily utilisable temperatures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
540390
(3,4-Dihydro-1-naphthyloxy)trimethylsilane, 97%
C13H18OSi