EMAIL THIS PAGE TO A FRIEND
Emergency nurse : the journal of the RCN Accident and Emergency Nursing Association
The properties of aminophylline.
PMID 18065044
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1755
Aminophylline, ≥98%, powder
C7H8N4O2 · 0.5C2H8N2