EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
Inhibition of matrix metalloproteinases prevents peroxynitrite-induced contractile dysfunction in the isolated cardiac myocyte.
PMID 18071296
Abstract
The potent oxidant peroxynitrite (ONOO(-)) induces mechanical dysfunction in the intact heart in part through activation of matrix metalloproteinase-2 (MMP-2). This effect may be independent of the proteolytic actions of MMPs on extracellular matrix proteins. The purpose of this study was to examine the effects of ONOO(-) on contractile function at the level of the single cardiac myocyte and whether this includes the action of MMPs. Freshly isolated ventricular myocytes from adult rats were superfused with Krebs-Henseleit buffer at 21 degrees C and paced at 0.5 Hz. Contractility was measured using a video edge-detector. ONOO(-) or decomposed ONOO(-) (vehicle control) were co-infused over 40 min to evaluate the contraction cease time (CCT). The effects of ONOO(-) on intracellular [Ca(2+)] were determined in myocytes loaded with calcium green-1 AM. MMP-2 activity was measured by gelatin zymography. ONOO(-) (30-600 microM) caused a concentration-dependent reduction in CCT. Myocytes subjected to 300 microM ONOO(-) had a shorter CCT than decomposed ONOO(-) (14.9+1.5 vs 32.2+3.5 min, n=7-8; P<0.05) and showed increased MMP-2 activity. The MMP inhibitors doxycycline (100 microM) or PD 166793 (2 microM) reduced the decline in CCT induced by 300 microM ONOO(-). ONOO(-) caused shorter calcium transient cease time and significant alterations in intracellular [Ca(2+)] homoeostasis which were partially prevented by doxycycline. This is the first demonstration that inhibition of MMPs protects the cardiac myocyte from ONOO(-)-induced contractile failure via an action unrelated to proteolysis of extracellular matrix proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0047
PD166793 hydrate, ≥98% (HPLC)
C17H18BrNO4S · xH2O