EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Synthesis and identification of propylene glycol phenyl ethers.
PMID 18102902
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
77766
1-Phenoxy-2-propanol, technical, ≥80% (GC)
C9H12O2