EMAIL THIS PAGE TO A FRIEND
Nature materials
High-capacity hydrogen storage in lithium and sodium amidoboranes.
PMID 18157135
Abstract
The safe and efficient storage of hydrogen is widely recognized as one of the key technological challenges in the transition towards a hydrogen-based energy economy. Whereas hydrogen for transportation applications is currently stored using cryogenics or high pressure, there is substantial research and development activity in the use of novel condensed-phase hydride materials. However, the multiple-target criteria accepted as necessary for the successful implementation of such stores have not yet been met by any single material. Ammonia borane, NH3BH3, is one of a number of condensed-phase compounds that have received significant attention because of its reported release of approximately 12 wt% hydrogen at moderate temperatures (approximately 150 degrees C). However, the hydrogen purity suffers from the release of trace quantities of borazine. Here, we report that the related alkali-metal amidoboranes, LiNH2BH3 and NaNH2BH3, release approximately 10.9 wt% and approximately 7.5 wt% hydrogen, respectively, at significantly lower temperatures (approximately 90 degrees C) with no borazine emission. The low-temperature release of a large amount of hydrogen is significant and provides the potential to fulfil many of the principal criteria required for an on-board hydrogen store.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
710199
Lithium ammonia borane, technical grade
H5BLiN