EMAIL THIS PAGE TO A FRIEND
The Journal of cell biology
DNA-activated protein kinase functions in a newly observed S phase checkpoint that links histone mRNA abundance with DNA replication.
PMID 18158334
Abstract
DNA and histone synthesis are coupled and ongoing replication is required to maintain histone gene expression. Here, we expose S phase-arrested cells to the kinase inhibitors caffeine and LY294002. This uncouples DNA replication from histone messenger RNA (mRNA) abundance, altering the efficiency of replication stress-induced histone mRNA down-regulation. Interference with caffeine-sensitive checkpoint kinases ataxia telangiectasia and Rad3 related (ATR)/ataxia telangiectasia mutated (ATM) does not affect histone mRNA down- regulation, which indicates that ATR/ATM alone cannot account for such coupling. LY294002 potentiates caffeine's ability to uncouple histone mRNA stabilization from replication only in cells containing functional DNA-activated protein kinase (DNA-PK), which indicates that DNA-PK is the target of LY294002. DNA-PK is activated during replication stress and DNA-PK signaling is enhanced when ATR/ATM signaling is abrogated. Histone mRNA decay does not require Chk1/Chk2. Replication stress induces phosphorylation of UPF1 but not hairpin[corrected]-binding protein/stem-loop binding protein at S/TQ sites, which are preferred substrate recognition motifs of phosphatidylinositol 3-kinase-like kinases, which indicates that histone mRNA stability may be directly controlled by ATR/ATM- and DNA-PK-mediated phosphorylation of UPF1.