EMAIL THIS PAGE TO A FRIEND
Foot & ankle international
Achilles tendon rupture associated with combination therapy of levofloxacin and steroid in four patients and a review of the literature.
PMID 18173993