EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Activity of a hydroxybibenzyl bryophyte constituent against Leishmania spp. and Trypanosoma cruzi: in silico, in vitro and in vivo activity studies.
PMID 18192088
Abstract
The synthesis and potent antiprotozoal activity of 14-hydroxylunularin, a natural hydroxybibenzyl bryophyte constituent is reported. 14-hydroxylunularin was highly active in vitro assays against culture and intracellular forms of Leishmania spp. and Trypanosoma. cruzi, in absence of cytotoxicity against mammalian cells. Preliminary structure-activity relationship studies showed that the reported bioactivity depends on hybridization at the carbon-carbon bridge, position and number of free hydroxy group on the aromatic rings. The reported results were also in agreement with the in silico prediction using Non-Stochastic Quadratic Fingerprints-based algorithms. The same compound also showed antiprotozoal activity in Leishmania spp. infected mice by oral and subcutaneous administration routes, with an optimal treatment of a daily subcutaneous administration of 10 mg/kg of body weight for 15 days. This study suggested that 14-hydroxylunularin may be chosen as a new candidate in the development of leishmanicidal therapy.