EMAIL THIS PAGE TO A FRIEND
Environmental health perspectives
Weight of the evidence on the human carcinogenicity of 2,4-D.
PMID 1820267
Abstract
The phenoxy herbicide 2,4-dichlorophenoxyacetic acid (2,4-D) is widely used to control the growth of weeds and broadleaf plants. We convened a panel of 13 scientists to weigh the evidence on the human carcinogenicity of 2,4-D. The panel based its findings on a review of the toxicological and epidemiological literature on 2,4-D and related phenoxy herbicides. The toxicological data do not provide a strong basis for predicting that 2,4-D is a human carcinogen. Although a cause-effect relationship is far from being established, the epidemiological evidence for an association between exposure to 2,4-D and non-Hodgkin's lymphoma is suggestive and requires further investigation. There is little evidence of an association between use of 2,4-D and soft-tissue sarcoma or Hodgkin's disease, and no evidence of an association between 2,4-D use and any other form of cancer. Scientists on the panel were asked to categorize 2,4-D as a "known," "probable," "possible," or "unlikely" carcinogen or as a noncarcinogen in humans. The predominant opinion among the panel members was that the weight of the evidence indicates that it is possible that exposure to 2,4-D can cause cancer in humans, although not all of the panelists believed the possibility was equally likely: one thought the possibility was strong, leaning toward probable, and five thought the possibility was remote, leaning toward unlikely. Two panelists believed it unlikely that 2,4-D can cause cancer in humans.