EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Pd-catalyzed aryl amination mediated by well defined, N-heterocyclic carbene (NHC)-Pd precatalysts, PEPPSI.
PMID 18224651
Abstract
Pd-N-heterocyclic carbene (NHC)-catalyzed Buchwald-Hartwig amination protocols mediated by Pd-PEPPSI precatalysts is described. These protocols provide access to a range of hindered and functionalized drug-like aryl amines in high yield with both electron-deficient and electron-rich aryl- and heteroaryl chlorides and bromides. Variations in solvent polarity, base and temperature are tolerated, enhancing the scope and utility of this protocol. A mechanistic rationalization for base strength (pKb) requirements is also provided.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
669032
PEPPSI-IPr catalyst, 98%
C32H40Cl3N3Pd
674702
PEPPSI-SIPr catalyst
C32H42Cl3N3Pd