EMAIL THIS PAGE TO A FRIEND
Journal of cell science
LUMA interacts with emerin and influences its distribution at the inner nuclear membrane.
PMID 18230648
Abstract
We present here a first characterization of LUMA, an unique integral inner nuclear membrane (INM) protein. LUMA is a highly conserved protein even in some bacteria and shares a PFAM domain of unknown function with orthologs from many species. Assessing LUMA topology by using protease protection of membrane-inserted LUMA and antibody epitope accessibility assays reveals that LUMA contains four transmembrane domains and a large hydrophilic domain located between membrane spans 1 and 2. The large hydrophilic domain is exposed to the perinuclear space whereas both LUMA termini reside cyto- or nucleoplasmically. Nuclear envelope targeting of LUMA mainly depends on the membrane spans. LUMA's transmembrane domains also promote homooligomerization. LUMA binds A- and B-type lamins and depends on A-type lamins for its INM localization. Furthermore, it interacts with emerin. Both downregulation of LUMA and overexpression of dominant-negative acting LUMA fragments causes redistribution of emerin. We propose that LUMA functions as a tetraspanin-like membrane organizer and has the potential to contribute to the pathomechanism of dystrophic diseases, such as Emery-Dreifuss muscular dystrophy.