EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Water-soluble 3-O-(2-methoxyethyl)cellulose: synthesis and characterization.
PMID 18261718
Abstract
The synthesis of novel 3-O-(2-methoxyethyl)cellulose via 2,6-di-O-thexyldimethylsilyl ethers was successfully carried out. Treatments of 3-O-(2-methoxyethyl)-2,6-di-O-thexyldimethylsilylcellulose with tetrabutylammonium fluoride trihydrate led to a complete removal of the protecting groups. Structure characterization carried out by means of 1D and 2D NMR spectroscopy proves a high regioselectivity. The novel cellulose ether is soluble in dimethyl sulfoxide, N,N-dimethylacetamide, N-methylpyrrolidone, and water. Size-exclusion chromatography revealed a distinct aggregation behavior in water.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
86843
Tetrabutylammonium fluoride trihydrate, technical, ≥90% (T)
C16H36FN · 3H2O