EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
DNA gyrase from the albicidin producer Xanthomonas albilineans has multiple-antibiotic-resistance and unusual enzymatic properties.
PMID 18268084
Abstract
The sugarcane pathogen Xanthomonas albilineans produces a family of antibiotics and phytotoxins termed albicidins, which inhibit plant and bacterial DNA gyrase supercoiling activity, with a 50% inhibitory concentration (50 nM) comparable to those of coumarins and quinolones. Here we show that X. albilineans has an unusual, antibiotic-resistant DNA gyrase. The X. albilineans gyrA and gyrB genes are not clustered with previously described albicidin biosynthesis and self-protection genes. The GyrA and GyrB products differ from Escherichia coli homologues through several insertions and through changes in several amino acid residues implicated in quinolone and coumarin resistance. Reconstituted X. albilineans DNA gyrase showed 20- to 25-fold-higher resistance than E. coli DNA gyrase to albicidin and ciprofloxacin and 8-fold-higher resistance to novobiocin in the supercoiling assay. The X. albilineans DNA gyrase is unusual in showing a high degree of distributive supercoiling and little DNA relaxation activity. X. albilineans GyrA (XaA) forms a functional gyrase heterotetramer with E. coli GyrB (EcB) and can account for albicidin and quinolone resistance and low levels of relaxation activity. XaB probably contributes to both coumarin resistance and the distributive supercoiling pattern. Although XaB shows fewer apparent changes relative to EcB, the EcA.XaB hybrid relaxed DNA in the presence or absence of ATP and was unable to supercoil. A fuller understanding of structural differences between albicidin-sensitive and -resistant gyrases may provide new clues into features of the enzyme amenable to interference by novel antibiotics.