EMAIL THIS PAGE TO A FRIEND
International journal of urology : official journal of the Japanese Urological Association
Mechanism of calcium oxalate renal stone formation and renal tubular cell injury.
PMID 18269444
Abstract
Formation of calcium oxalate stones tends to increase with age and begins from the attachment of a crystal formed in the cavity of renal tubules to the surface of renal tubular epithelial cells. Though most of the crystals formed in the cavity of renal tubules are discharged as is in the urine, in healthy people, crystals that attach to the surface of renal tubular epithelial cells are thought to be digested by macrophages and/or lysosomes inside of cells. However, in individuals with hyperoxaluria or crystal urine, renal tubular cells are injured and crystals easily become attached to them. Various factors are thought to be involved in renal tubular cell injury. Crystals attached to the surface of renal tubular cells are taken into the cells (crystal-cell interaction). And then the crystal and crystal aggregates grow, and finally a stone is formed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
455997
Calcium oxalate, ≥99.9% trace metals basis
C2CaO4