EMAIL THIS PAGE TO A FRIEND
Journal of dermatological science
Histidase expression in human epidermal keratinocytes: regulation by differentiation status and all-trans retinoic acid.
PMID 18280705
Abstract
Histidase (histidine ammonia lyase) converts histidine into urocanic acid, the main ultraviolet (UV) light absorption factor of the stratum corneum. It is unknown if and how histidase is regulated in the epidermis. We have investigated the transcriptional regulation of histidase expression in epidermal keratinocytes. Human epidermal keratinocytes were cultured in vitro and exposed to UV irradiation, a number of cytokines and all-trans retinoic acid (ATRA) (1 microM). Keratinocyte differentiation was triggered by maintaining confluent cells in monolayer culture and by establishing three-dimensional skin equivalents. The mRNA expression level of histidase in keratinoytes as well as in the epidermis and other tissues was determined by quantitative real-time PCR. Protein expression was determined by Western blot analysis. Human epidermis contained higher levels of histidase transcripts than all other tissues investigated. Expression of histidase strongly increased at the mRNA and protein levels during differentiation of primary keratinocytes in vitro. Treatment of keratinocytes with UVA and UVB did not significantly change the expression level of histidase. By contrast, ATRA suppressed histidase expression almost completely. Our results show that histidase is upregulated during keratinocyte differentiation and that ATRA but not UV irradiation modulates the expression level of histidase. Suppression of histidase-mediated production of urocanic acid may contribute to the increase in UV sensitivity that is caused by treatment with retinoids.