EMAIL THIS PAGE TO A FRIEND
Zhongguo dang dai er ke za zhi = Chinese journal of contemporary pediatrics
[Mechanism and clinical application of erythromycin as a gastrointestinal prokinetic agent in children].
PMID 18289489