EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis, antimicrobial activity, pharmacophore analysis of some new 2-(substitutedphenyl/benzyl)-5-[(2-benzofuryl)carboxamido]benzoxazoles.
PMID 18289735
Abstract
The synthesis and antimicrobial activity of a new series of 2-(substitutedphenyl/benzyl)-5-[(2-benzofuryl)carboxamido]benzoxazole derivatives 3-12 were described. The in vitro antimicrobial activity of the compounds was determined against some Gram-positive, Gram-negative bacteria and fungi and their drug-resistant isolates in comparison with standard drugs. Antimicrobial results indicated that the synthesized compounds possessed a broad spectrum of activity with MIC values 500-15.625 microg/ml. In the series, the most active compound against Candida krusei and Candida albicans isolate is 8 with MIC value 31.25 microg/ml. However, it is one dilution less potent than the compared fluconazole. Some of the screened compounds exhibit significant activity, having MIC value as 31.25 microg/ml in Pseudomonas aeruginosa having same activity as Rifampicin. Furthermore, considering the worth of developing new antibacterial agents against drug-resistant P. aeruginosa the present study explores the structure-activity relationship analysis of 2-(substitutedphenyl/benzyl)-5-[(2-benzofuryl)carboxamido]benzoxazoles using 3D-common features pharmacophore hypotheses approach.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A6140
Ampicillin trihydrate, 96.0-100.5% anhydrous basis (HPLC)
C16H19N3O4S · 3H2O
31591
Ampicillin trihydrate, VETRANAL, analytical standard
C16H19N3O4S · 3H2O