EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Is vanadium a more versatile target in the activity of primordial life forms than hitherto anticipated?
PMID 18327316
Abstract
The two predominant forms of vanadium occurring in the geo-, aqua- and biosphere, soluble vanadate(V) and insoluble oxovanadium(IV) (vanadyl), are subject to bacterial activity and transformation. Bacteria belonging to genera such as Shewanella, Pseudomonas and Geobacter can use vanadate as a primary electron acceptor in dissimilation or respiration, an important issue in the context of biomineralisation and soil detoxification. Azotobacter, which can employ vanadium as an essential element in nitrogen fixation, secretes a vanadophore which enables the uptake of vanadium(V). Siderophores secreted by other bacteria competitively (to ferric iron) take up vanadyl and thus interfere with iron supply, resulting in bacteriostasis. The halo-alkaliphilic Thioalkalivibrio nitratireducens possibly uses vanadium as a constituent of an alternative, molybdopterin-free nitrate reductase. Marine macro-algae can generate a variety of halogenated organic compounds by use of vanadate-dependent haloperoxidases, and a molecular vanadium compound, amavadin, from Amanita mushrooms has turned out to be an efficient catalyst in oxidation reactions. The present account is a focused and critical review of the current knowledge of the interplay of bacteria and other primitive forms of life (cyanobacteria, algae, fungi and lichens) with vanadium, with the aim to provide perspectives for applications and further investigations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF98785388
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF25800386
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
V
GF46548692
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF21697106
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF76957004
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF23355996
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF69763977
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
V
GF75438445
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
V
GF84027269
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
V
GF25047503
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
V
GF39723239
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
V
GF29805341
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
V
GF03408555
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
V
GF30937732
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
V
GF57656391
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
V
GF25668344
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
V
GF26068855
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
V
GF67217430
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
V
GF78288946
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
V
GF08542975
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
V
GF58606409
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
V
GF02179672
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
V
GF88432432
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%
V
GF67872213
Vanadium, wire reel, 5m, diameter 0.25mm, hard, 99.8%
V
GF51153724
Vanadium, microfoil, disks, 10mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
V