EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Targeting gastrin-releasing peptide receptors of prostate cancer cells for photodynamic therapy with a phthalocyanine-bombesin conjugate.
PMID 18329268
Abstract
Sulfonated aluminum phthalocyanines (AlPcS) are potent photosensitizers for the photodynamic therapy (PDT) of cancer. In this study we evaluate the possibility to improve the efficacy of AlPcS-PDT for prostate cancer by targeting tetrasulfonated aluminum phthalocyanines (AlPcS(4)) to the gastrin-releasing peptide receptor (GRPR) through coupling to bombesin. A mono-carbohexyl derivative of AlPcS(4) is attached to 8-Aoc-bombesin(7-14)NH(2) via an amide bridge to yield a bombesin-AlPcS(4) conjugate linked by a C-14 spacer chain. The conjugate is characterized by mass spectroscopy and shown to bind to the GRPR with a relative binding affinity (RBA) of 2.3, taking bombesin (RBA=100) as unity. The in vitro photodynamic efficacy of the conjugate against PC-3 human prostate cancer cells is improved by a factor 2.5 over the non-conjugated mono-carbohexyl derivative of AlPcS(4).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4272
Bombesin acetate salt hydrate, ≥97% (HPLC)
C71H110N24O18S · xC2H4O2 · yH2O