EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Kardiologie
[Pharmacology and pharmacokinetics of enoximone].
PMID 1833893
Abstract
Enoximone belongs to the imidazole class of compounds, that possess positive inotropic and vasodilatory activities. These pharmacologic effects are caused by selective inhibition of a cAMP-specific phosphodiesterase in the heart and in the smooth muscle of blood vessels. Results obtained in intact animals showed a dose-dependent increase in cardiac contractile force and a reduction in peripheral arterial resistance with only a slight increase in heart rate. Following acute administration of enoximone to patients suffering from cardiac failure an almost linear increase of cardiac index with increasing doses was found. Enoximone is eliminated from the body by biotransformation. Sulfoxide formation is the main metabolic step in man. This metabolite is excreted in the urine. Enoximone sulfoxide also possesses some cardiotonic activity. Reconversion of enoximone sulfoxide to enoximone was shown to occur in the liver and, to some extent, also in the kidney. Bioavailability of enoximone after a single oral dose of 3 mg/kg is about 55%, but may be higher following chronic therapy. This is probably due to saturation of the first-pass metabolism. A mean clearance of about 10 ml/min/kg and a mean half-life of 6 h were determined in patients with cardiac failure. These values are different from those measured in normal volunteers, indicating a reduced clearance of enoximone in these patients. In patients with renal failure enoximone sulfoxide accumulates in plasma. The elimination of enoximone is also reduced which might be caused by reconversion of enoximone sulfoxide to enoximone in this pathophysiologic condition. Thus, the dose of enoximone should be reduced in patients with severe impairment of renal function. Clinically significant drug interactions have not been reported so far.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1279
Enoximone, solid
C12H12N2O2S