EMAIL THIS PAGE TO A FRIEND
Journal of animal science
Use of 25-hydroxyvitamin D3 and dietary calcium to improve tenderness of beef from the round of beef cows.
PMID 18344286
Abstract
The objective of this trial was to determine how 25-hydroxyvitamin D(3) (25-OH D(3)) supplementation, altering supplemental dietary calcium, or their combination influence postmortem biochemical and tenderness changes in muscles from the round of mature cows. Twenty-seven Angus cows (3 to 7 yr old) were allotted randomly to 9 pens with 3 cows per pen. Treatments were arranged in a 3 x 3 factorial design with 3 dosages of 25-OH D(3) (0, 250, or 500 mg of 25-OH D(3) administered as a 1-time oral bolus 7 d before slaughter) and 3 percentages of supplemental limestone (0.5, 0.75, and 1.0%) replenished in the diet for 3 d before slaughter and after a 2-wk limestone withdrawal. Plasma samples were obtained during the feeding period. Upon slaughter, adductor, gracilus, pectineus, sartorius, semimembranosus, vastus intermedius, and vastus lateralis muscles were obtained and aged for 1, 3, or 7 d. Calcium concentrations were increased in plasma when 250 or 500 mg of 25-OH D(3) were administered (P
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17937
25-Hydroxyvitamin D2, ≥98.0% (HPLC)
C28H44O2