EMAIL THIS PAGE TO A FRIEND
Nature genetics
A mutation of human cytochrome c enhances the intrinsic apoptotic pathway but causes only thrombocytopenia.
PMID 18345000
Abstract
We report the first identified mutation in the gene encoding human cytochrome c (CYCS). Glycine 41, invariant throughout eukaryotes, is substituted by serine in a family with autosomal dominant thrombocytopenia caused by dysregulated platelet formation. The mutation yields a cytochrome c variant with enhanced apoptotic activity in vitro. Notably, the family has no other phenotypic indication of abnormal apoptosis, implying that cytochrome c activity is not a critical regulator of most physiological apoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2506
Cytochrome c from equine heart, ≥95% (SDS-PAGE)