EMAIL THIS PAGE TO A FRIEND
Nature
Molecular cloning and characterization of the rat NMDA receptor.
PMID 1834949
Abstract
A complementary DNA encoding the rat NMDA receptor has been cloned and characterized. The single protein encoded by the cDNA forms a receptor-channel complex that has electrophysiological and pharmacological properties characteristic of the NMDA receptor. This protein has a significant sequence similarity to the AMPA/kainate receptors and contains four putative transmembrane segments following a large extracellular domain. The NMDA receptor messenger RNA is expressed in neuronal cells throughout the brain regions, particularly in the hippocampus, cerebral cortex and cerebellum.