EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Rational design, synthesis, and in vivo evaluation of the antileukemic activity of six new alkylating steroidal esters.
PMID 18353651
Abstract
The synthesis and the in vivo evaluation against leukemias P388 and L1210 of six new alkylating steroidal esters are described. The esteric derivatives incorporating the 17beta-acetamido-B-lactamic steroidal skeleton exhibited increased antileukemic activity and lower toxicity, compared to the 17beta-acetamido-7-keto analogs. Among the 17beta-acetamido-B-lactamic steroidal esters, the most potent compound afforded four out of six cures in leukemia P388 and was measured to be almost non-toxic, producing significant low levels of toxicity.