EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Mechanisms of macrolide resistance among Streptococcus pneumoniae isolates from Russia.
PMID 18378707
Abstract
Among 76 macrolide-nonsusceptible Streptococcus pneumoniae isolates collected between 2003 and 2005 from Central Russia, the resistance mechanisms detected in the isolates included erm(B) alone (50%), mef alone [mef(E), mef(I), or a different mef subclass; 19.7%], or both erm(B) and mef(E) (30.3%). Isolates with dual resistance genes [erm(B) and mef(E)] belonged to clonal complex CC81 or CC271.