EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Transcriptional regulation of chondrogenesis by coactivator Tip60 via chromatin association with Sox9 and Sox5.
PMID 18390577
Abstract
Sox9 is a transcription factor of the SRY family required for several steps of chondrogenesis. It activates the expression of various chondrocyte-specific genes, but the mechanisms and role of cofactors involved in Sox9-regulated gene transcription are not fully understood. Here, we report on the characterization of a Tat interactive protein-60 (Tip60) as Sox9-associated protein identified in a yeast two-hybrid screen. Both in vitro and in vivo assays confirmed the specificity of interactions between Sox9 and Tip60 including the existence of an endogenous complex containing both polypeptides in chondrocytes. Gel shift assays showed the presence of a complex containing Sox9, Tip60 and the DNA of an enhancer region of the Col2a1 promoter. Reporter assays using a Col2a1 promoter with multimerized Col2a1 Sox9-binding sites indicated that Tip60 enhanced the transcriptional activity of Sox9. A larger Col2a1 promoter showed that Tip60 increased the activity of this promoter in the presence of both Sox9 and Sox5. Ectopic expression of Sox9 and transient-cotransfection with Tip60 in COS7 cells showed a more diffuse subnuclear colocalization, suggesting changes in the chromatin structure. Chromatin immunoprecipitation assays showed that Tip60, Sox9 and Sox5 associated with the same Col2a1 enhancer region. Consistent with a role of Tip60 in chondrogenesis, addition of Tip60 siRNA to limb-bud micromass cultures delayed chondrocyte differention. Tip60 enhances acetylation of Sox9 mainly through K61, 253, 398 residues; however, the K61/253/398A mutant of Sox9 still exhibited enhanced transcriptional activity by Tip60. Our results support the hypothesis that Tip60 is a coactivator of Sox9 in chondrocytes.