EMAIL THIS PAGE TO A FRIEND
Cancer research
MUC1 expression is regulated by DNA methylation and histone H3 lysine 9 modification in cancer cells.
PMID 18413738
Abstract
MUC1 is a transmembrane mucin that is highly expressed in various cancers and correlates with malignant potential. Important cancer-related genes such as p16 and E-cadherin are controlled epigenetically; however, MUC1 has been overlooked in epigenetics. Herein, we provide the first report that MUC1 gene expression is regulated by DNA methylation and histone H3 lysine 9 (H3-K9) modification of the MUC1 promoter. The recently developed MassARRAY assay was performed to investigate the DNA methylation status of 184 CpG sites from -2,753 to +263. Near the transcriptional start site, the DNA methylation level of MUC1-negative cancer cell lines (e.g., MDA-MB-453) was high, whereas that of MUC1-positive cell lines (e.g., MCF-7) was low. Histone H3-K9 modification status was also closely related to MUC1 gene expression. Furthermore, MUC1 mRNA expression in MUC1-negative cells was restored by treatment with the DNA methylation inhibitor 5-aza-2'-deoxycytidine. Our results indicate that DNA methylation and histone H3-K9 modification in the 5' flanking region play a critical role in MUC1 gene expression, and this study defines MUC1 as a new member of the class of epigenetically controlled genes. An understanding of the epigenetic changes of MUC1 may be of importance for diagnosis of carcinogenic risk and prediction of outcome for cancer patients.