EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong xi yi jie he za zhi Zhongguo Zhongxiyi jiehe zazhi = Chinese journal of integrated traditional and Western medicine
[Experimental advance of applying emodin for prevention and treatment of liver diseases].
PMID 18418981
Abstract
The experimental researches of applying emodin for prevention and treatment of liver diseases in recent years were reviewed. Emodin can inhibit the growth of liver tumor cells in vitro and in vivo, inducing cell apoptosis is one of its mechanisms. Emodin also has the effects of liver protection, anti-liver fibrosis, and so on, the mechanisms for those effects still need more studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E7881
Emodin, from Frangula bark, ≥90% (HPLC)
C15H10O5
30269
Emodin, analytical standard
C15H10O5