EMAIL THIS PAGE TO A FRIEND
Diabetes
Improved vascular engraftment and graft function after inhibition of the angiostatic factor thrombospondin-1 in mouse pancreatic islets.
PMID 18420490
Abstract
Insufficient development of a new intra-islet capillary network after transplantation may be one contributing factor to the failure of islet grafts in clinical transplantation. The present study tested the hypothesis that the angiostatic factor thrombospondin-1 (TSP-1), which is normally present in islets, restricts intra-islet vascular expansion posttransplantation. Pancreatic islets of TSP-1-deficient (TSP-1(-/-)) mice or wild-type islets transfected with siRNA for TSP-1 were transplanted beneath the renal capsule of syngeneic or immunocompromised recipient mice. Both genetically TSP-1(-/-) islets and TSP-1 siRNA-transfected islet cells demonstrated an increased vascular density when compared with control islets 1 month after transplantation. This was also reflected in a markedly increased blood perfusion and oxygenation of the grafts. The functional importance of the improved vascular engraftment was analyzed by comparing glucose-stimulated insulin release from islet cells transfected with either TSP-1 siRNA or scramble siRNA before implantation. These experiments showed that the increased revascularization of grafts composed of TSP-1 siRNA-transfected islet cells correlated to increments in both their first and second phase of glucose-stimulated insulin secretion. Our findings demonstrate that inhibition of TSP-1 in islets intended for transplantation may be a feasible strategy to improve islet graft revascularization and function.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R7647 Random Nonamers, for use as primers in cDNA synthesis, 50 μM in H2O
R5886 RPMI-1640 Medium, HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture
R7388 RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
51536C RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture
R0883 RPMI-1640 Medium, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
R7638 RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
R1145 RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
R7513 RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
R7509 RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
R7755 RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
R4130 RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
R1383 RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
R8755 RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
R6504 RPMI-1640 Medium, With L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
R8005 RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
R1780 RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
R2405 RPMI-1640 Medium, AQmedia, With L-alanyl-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture