EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Antibacterial activity in serum of the 3,5-diamino-piperidine translation inhibitors.
PMID 18440814
Abstract
Translation inhibitors of the 3,5-diamino-piperidine series act as aminoglycoside mimetics that inhibit bacterial growth. Here we show antibacterial SAR in the presence and absence of serum with a particular focus toward Pseudomonas aeruginosa.