EMAIL THIS PAGE TO A FRIEND
Nutrition and cancer
Impact of lactose containing foods and the genetics of lactase on diseases: an analytical review of population data.
PMID 18444163
Abstract
Dairy foods (DFs) contain complex ingredients that could affect different diseases. The control of lactose digestion phenotypically divides populations into those who can [lactase persistent (LP)] and those who cannot [lactase nonpersistent (LNP)] assimilate lactose. LNP subjects, however, can adapt to lactose intolerance through intestinal bacteria. The DF/LNP status interactions may function as disease risk modifiers. We evaluated the relationship between DF and LNP with colorectal, breast, prostate, ovarian, lung, and stomach cancer and inflammatory bowel diseases (IBD; Crohn's disease and ulcerative colitis). Yearly per capita DF consumption, LNP national prevalence, cancer mortality, and incidence of IBD were obtained from several sources. A negative binomial regression model was used to derive incremental risks. There were statistically significant (P