EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Effects of amide constituents from pepper on adipogenesis in 3T3-L1 cells.
PMID 18477507
Abstract
Several amide constituents (piperlonguminine and retrofractamides A, B, and C) from the fruit of Piper chaba promoted adipogenesis of 3T3-L1 cells. Among them, retrofractamide A was the most active and significantly increased the amount of adiponectin released into the medium and the uptake of 2-deoxyglucose into the cells. Retrofractamide A also increased mRNA levels of adiponectin, peroxisome proliferator-activated receptor gamma2 (PPARgamma2), glucose transporter 4 (GLUT4), and insulin receptor substrate 1 (IRS-1), but did not act as a PPARgamma agonist different from troglitazone.