EMAIL THIS PAGE TO A FRIEND
Clinical interventions in aging
Carvedilol in the treatment of elderly patients with chronic heart failure.
PMID 18488879
Abstract
Chronic heart failure (CHF) is common, and increases in incidence and prevalence with age. There are compelling data demonstrating reduced mortality and hospitalizations with adrenergic blockade in older patients with CHF. Despite this, many older patients remain undertreated. The aim of the present article is to review the potential mechanisms of the benefits of adrenergic blockade in CHF and the clinical data available from the large randomized studies, focusing particularly on older patients.