EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
Synthesis and antimicrobial activities of some new triazolothiadiazoles bearing 4-methylthiobenzyl moiety.
PMID 18508161
Abstract
A series of substituted triazolothiadiazoles have been synthesized by condensing 4-amino-3-[4-methylthiobenzyl]-4H-1,2,4-triazole-5-thiol (5) with substituted aryl furoic acids/aromatic acids in the presence of POCl3. The triazole (5) was obtained by the fusion of 4-methylthiophenyl acetic acid with thiocarbohydrazide. The structures of newly synthesized compounds are characterized by elemental analysis, IR, 1H NMR and mass spectroscopic studies and were screened for their antimicrobial activities. The preliminary results revealed that some of the compounds exhibited promising antimicrobial activities.