EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Discovery of nonsteroidal glucocorticoid receptor ligands based on 6-indole-1,2,3,4-tetrahydroquinolines.
PMID 18513967
Abstract
A series of nonsteroidal glucocorticoid receptor (GR) ligands based on a 6-indole-1,2,3,4-tetrahydroquinoline scaffold are reported. Structure-activity relationship (SAR) of the pendent indole group identified compound 20 exhibiting good GR binding affinity (K(i)=1.5nM) and 100- to 1000-fold selectivity over MR, PR, and AR while showing activity in an E-selectin repression assay.