EMAIL THIS PAGE TO A FRIEND
Trends in pharmacological sciences
Regulation of AMPA receptor gating and pharmacology by TARP auxiliary subunits.
PMID 18514334
Abstract
Presynaptic glutamate release elicits brief waves of membrane depolarization in neurons by activating AMPA receptors. Depending on its precise size and shape, current through AMPA receptors gates downstream processes like NMDA receptor activation and action potential generation. Over a decade of research on AMPA receptor structure and function has identified binding sites on AMPA receptors for agonists, antagonists and allosteric modulators as well as key residues underlying differences in the gating behavior of various AMPA receptor subtypes. However, the recent discovery that AMPA receptors are accompanied in the synaptic membrane by a family of auxiliary subunits known as transmembrane AMPA receptor regulatory proteins (TARPs) has revealed that the kinetics and pharmacology of neuronal AMPA receptors differ in many respects from those predicted by classical studies of AMPA receptors in heterologous systems. Here, we summarize recent work and discuss remaining questions concerning the structure and function of native TARP-AMPA receptor complexes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G111
γ-D-Glutamylaminomethylsulfonic acid, solid
C6H12N2O6S