EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Improved biodesulfurization of hydrodesulfurized diesel oil using Rhodococcus erythropolis and Gordonia sp.
PMID 18516503
Abstract
Substituted benzothiophenes (BTs) and dibenzothiophenes (DBTs) remain in diesel oil following conventional desulfurization by hydrodesulfurization. A mixture of washed cells (13.6 g dry cell wt l(-1)) of Rhodococcus erythropolis DS-3 and Gordonia sp. C-6 were employed to desulfurize hydrodesulfurized diesel oil; its sulfur content was reduced from 1.26 g l(-1) to 180 mg l(-1), approx 86% (w/w) of the total sulfur was removed from diesel oil after three cycles of biodesulfurization. The average desulfurization rate was 0.22 mg sulfur (g dry cell wt)(-1) h(-1). A bacterial mixture is therefore efficient for the practical biodesulfurization of diesel oil.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
467464
Thianaphthene-2-carboxylic acid, 98%
C9H6O2S