EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
Spontaneous development of a pancreatic exocrine disease in CD28-deficient NOD mice.
PMID 18523243
Abstract
Autoimmune pancreatitis (AIP) is a heterogeneous autoimmune disease in humans characterized by a progressive lymphocytic and plasmacytic infiltrate in the exocrine pancreas. In this study, we report that regulatory T cell-deficient NOD.CD28KO mice spontaneously develop AIP that closely resembles the human disease. NOD mouse AIP was associated with severe periductal and parenchymal inflammation of the exocrine pancreas by CD4(+) T cells, CD8(+) T cells, and B cells. Spleen CD4(+) T cells were found to be both necessary and sufficient for the development of AIP. Autoantibodies and autoreactive T cells from affected mice recognized a approximately 50-kDa protein identified as pancreatic amylase. Importantly, administration of tolerogenic amylase-coupled fixed spleen cells significantly ameliorated disease severity, suggesting that this protein functions as a key autoantigen. The establishment and characterization of this spontaneous pancreatic amylase-specific AIP in regulatory T cell-deficient NOD.CD28KO mice provides an excellent model for the study of disease pathogenesis and development of new therapies for human autoimmune pancreatitis.