EMAIL THIS PAGE TO A FRIEND
European journal of medicinal chemistry
A class of potent tyrosinase inhibitors: alkylidenethiosemicarbazide compounds.
PMID 18524420
Abstract
A series of alkylidenethiosemicarbazide compounds were synthesized and their inhibitory effects on the diphenolase activity of mushroom tyrosinase were evaluated. The results showed that most of the synthesized compounds exhibited significant inhibitory activities. Especially, compound 1f was found to be the most potent inhibitor with IC(50) value of 0.086 microM, suggesting that further development of such compounds may be of interest.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M13807
4-Methoxycinnamic acid, predominantly trans, 99%
C10H10O3
T89702
Tropolone, 98%
C7H6O2